Learn More
Abcam Inosine, Purine nucleoside, 1G
Catalog No. 502707333
Encompass_Preferred
MW 268.23 g/mol, Purity >99%. Purine nucleoside and adenosine metabolite. Cell-permeable. Displays neuroprotective and immunomodulatory effects. Potent vasodilator and axon growth stimulator.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7333 Supplier Abcam Supplier No. AB1207241G
Add to Cart
Add to Cart
MW 268.23 g/mol, Purity >99%. Purine nucleoside and adenosine metabolite. Cell-permeable. Displays neuroprotective and immunomodulatory effects. Potent vasodilator and axon growth stimulator.
The product is subject to the following: Abcam Restricted Use Statement