Learn More
Medchemexpress LLC Inosinic acid | 131-99-7 | MFCD30342155 | 99.9% | 348.21 g/mol | C10H13N4O8P | 100 MG
SDP
Catalog No. 5000241714
Encompass_Preferred
Inosinic acid (inosine 5'-monophosphate, IMP) is an endogenous nucleotide metabolite supplied as a purified research-grade powder. It is used in flavor research as an umami tastant, in animal nutrition studies to assess effects on growth and meat quality, and in biochemical assays involving nucleotide metabolism.
  • Purified compound for analytical and formulation studies
  • High reported purity (≈99.9%)
  • Molecular formula C10H13N4O8P and molecular weight 348.21 g/mol
  • Water-soluble nucleotide suitable for biochemical and food science assays
  • Available in small mg-scale packages for laboratory use
Catalog No. 50-002-41714 Supplier Medchemexpress LLC Supplier No. HY108213100MG
Add to Cart
Add to Cart
Inosinic acid (inosine 5'-monophosphate, IMP) is an endogenous nucleotide metabolite supplied as a purified research-grade powder. It is used in flavor research as an umami tastant, in animal nutrition studies to assess effects on growth and meat quality, and in biochemical assays involving nucleotide metabolism.
  • Purified compound for analytical and formulation studies
  • High reported purity (≈99.9%)
  • Molecular formula C10H13N4O8P and molecular weight 348.21 g/mol
  • Water-soluble nucleotide suitable for biochemical and food science assays
  • Available in small mg-scale packages for laboratory use