Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inositol, 97%, Spectrum™ Chemical
SDP

Catalog No. 18611039 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-039 100 g Poly Bottle
18-611-043 500 g Poly Bottle
18-611-041 2.5 kg Poly Bottle
18-611-040 12 kg Poly Bottle
18-611-042 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-039 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IN140100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

CAS Max % 1
Melting Point 224° to 227°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.97
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Inositol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.