Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inositol, FCC, 97%, Spectrum™ Chemical
SDP

Catalog No. 18603447 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-447 100 g Poly Bottle
18-603-448 500 g Poly Bottle
18-603-449 2.5 kg Poly Bottle
18-603-451 25 kg Poly Bottle
18-603-450 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-447 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I1004100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

I1004, 87-89-8, C6H12O6

Spectrum™ Chemical Inositol, FCC is a vitamin-like substance, sometimes known colloquially asvitamin B-8, is a beneficial nutrient that has been implicated in the treatment of some behavioral or emotional disorders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES OC1C(O)C(O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 224-227 C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 97%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Inositol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.