Learn More
Echelon Biosciences Research Labs Ins (1,3,4,5)P4 Beads 1 mL
SDP
Catalog No. 50720747
Encompass
Ins(1,3,4,5)P4 Beads are composed of agarose with 10 nanomoles of bound Ins(1,3,4,5)P4 per mL of beads, enough for several protein binding experiments. Each 1 mL order of Ins(1,3,4,5)P4 Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-747 Supplier Echelon Biosciences Research Labs Supplier No. QB13451ML
Ins(1,3,4,5)P4 Beads are composed of agarose with 10 nanomoles of bound Ins(1,3,4,5)P4 per mL of beads, enough for several protein binding experiments. Each 1 mL order of Ins(1,3,4,5)P4 Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable