Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inositol nicotinate | 6556-11-2 | 98.1% | 25 MG
SDP

Catalog No. 5000231255
Encompass_Preferred

Inositol nicotinate is the myo-inositol ester of nicotinic acid used in research to study vasodilation and peripheral arterial disease. It is supplied in defined small pack sizes for laboratory use and is characterized by high purity and a defined molecular weight.

  • High purity: 98.1%.
  • Molecular weight: 810.72 g/mol.
  • Chemical formula: C42H30N6O12.
  • Available pack sizes: 25 MG, 50 MG, 100 MG, 200 MG, 500 MG.
  • Intended for research use only.

Catalog No. 50-002-31255 Supplier Medchemexpress LLC Supplier No. HY12236525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inositol nicotinate is the myo-inositol ester of nicotinic acid used in research to study vasodilation and peripheral arterial disease. It is supplied in defined small pack sizes for laboratory use and is characterized by high purity and a defined molecular weight.

  • High purity: 98.1%.
  • Molecular weight: 810.72 g/mol.
  • Chemical formula: C42H30N6O12.
  • Available pack sizes: 25 MG, 50 MG, 100 MG, 200 MG, 500 MG.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.