Learn More
Crescent Chemical Co Inc INOSITOL RESEARCH GRADE
SDP
Catalog No. 50247745
Encompass_Preferred
Inositol research grade, USP/NF For bacteriology. Optically inactive. Tested for its suitability in tissue culture. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 26310.01 CAS 87-89-8
Catalog No. 50-247-745 Supplier Crescent Chemical Co Inc Supplier No. SE26310.01
Inositol research grade, USP/NF For bacteriology. Optically inactive. Tested for its suitability in tissue culture. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 26310.01 CAS 87-89-8