Learn More
TRIBIOSCIENCE INC Inoue Transformation Buffer, 100ML
SDP
Catalog No. NC3523891
Encompass
Inoue Transformation Buffer is used for the preparation of high-efficient competent cells. This product is based on the Inoue method for the preparation and transformation of E. Coli component cells.
Catalog No. NC3523891 Supplier TRIBIOSCIENCE INC Supplier No. TBS8036100ML
Inoue Transformation Buffer is used for the preparation of high-efficient competent cells. This product is based on the Inoue method for the preparation and transformation of E. Coli component cells.