Learn More
TRIBIOSCIENCE INC Inoue Transformation Buffer, 500ML
SDP
Catalog No. NC3523892
Encompass
Inoue Transformation Buffer is used for the preparation of high-efficient competent cells. This product is based on the Inoue method for the preparation and transformation of E. Coli component cells.
Catalog No. NC3523892 Supplier TRIBIOSCIENCE INC Supplier No. TBS8036500ML
Inoue Transformation Buffer is used for the preparation of high-efficient competent cells. This product is based on the Inoue method for the preparation and transformation of E. Coli component cells.