Learn More
Ace Glass, Inc. Stopper, impinger, tube end inlet/outlets, 24/40, calibrated nozzle
SDP
Catalog No. NC0591096
Encompass_Preferred
24/40 midget impinger stopper with tube end inlet/outlets, and a calibrated nozzle that delivers 0.09-0.11 CFM at 30.5cm vacuum.
Catalog No. NC0591096 Supplier Ace Glass, Inc. Supplier No. 753106
24/40 midget impinger stopper with tube end inlet/outlets, and a calibrated nozzle that delivers 0.09-0.11 CFM at 30.5cm vacuum.