Learn More
Aviva Systems Biology Corp INPP5D Antibody - C-terminal region (ARP61033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INPP5D. It was validated on Western Blot. . Alternate names: MGC104855, MGC142140, MGC142142, SHIP, SHIP1, SIP-145, hp51CNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-235