Learn More
Aviva Systems Biology Corp INPP5K Antibody - N-terminal region (ARP61153_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61153_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INPP5K. It was validated on Western Blot. . Alternate names: PPS, SKIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-275