Learn More
Aviva Systems Biology Corp INPP5K Antibody - C-terminal region (ARP61152_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61152_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INPP5K. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human INPP5K. Alternate names: PPS, SKIPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-274