Learn More
METTLER TOLEDO pH Sensor InPro 3100i SG/120
Supplier:  METTLER TOLEDO 52003515
Encompass
The InPro 3100i SG/120 is a combined pH/ORP electrode and temperature sensor designed for measurement in bioprocesses where CIP and SIP procedures are required. The 120 mm version is optimal for short installation lengths.
Catalog No. NC0785640