Learn More
METTLER TOLEDO pH Sensor InPro 3250 225/PT1000
Supplier:  METTLER TOLEDO 52002553
Encompass
The InPro 3250 225/PT1000 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.
Catalog No. NC0482522