Learn More
METTLER TOLEDO pH Sensor InPro 3250i SG/120
Supplier:  METTLER TOLEDO 52005373
Encompass
The InPro 3250i SG/120 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.
Catalog No. NC9938760