Learn More
METTLER TOLEDO pH Sensor InPro3250i/SG/325
Catalog No. NC0744063 Shop All METTLER TOLEDO Products
Encompass
The InPro 3250i SG/325 is a digital, pre-pressurized, low maintenance ISM pH & ORP sensor combined with a temperature sensor for in-line measurement in demanding applications. The 325 mm version is optimal for medium to long installation lengths.
Catalog No. NC0744063 Supplier METTLER TOLEDO Supplier No. 52005375
The InPro 3250i SG/325 is a digital, pre-pressurized, low maintenance ISM pH & ORP sensor combined with a temperature sensor for in-line measurement in demanding applications. The 325 mm version is optimal for medium to long installation lengths.