Learn More
METTLER TOLEDO pH Sensor InPro 3253 120/PT1000
Catalog No. NC0024970 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253 120/PT1000 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.
Catalog No. NC0024970 Supplier METTLER TOLEDO Supplier No. 52002567
The InPro 3253 120/PT1000 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.