Learn More
METTLER TOLEDO pH Sensor InPro 3253 325/PT100
Catalog No. NC0341716 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253 325/PT100 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. The 325 mm version is optimal for medium installation lengths.
Catalog No. NC0341716 Supplier METTLER TOLEDO Supplier No. 52002571
The InPro 3253 325/PT100 is a pre-pressurized, liquid filled, low maintenance pH sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. The 325 mm version is optimal for medium installation lengths.