Learn More
METTLER TOLEDO pH Sensor InPro3253/120/PT100
Catalog No. NC9235027 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253 120/PT100 is a pre-pressurized, liquid filled, low maintenance pH sensor combined with a PT100 temperature sensor for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.
Catalog No. NC9235027 Supplier METTLER TOLEDO Supplier No. 52002566
The InPro 3253 120/PT100 is a pre-pressurized, liquid filled, low maintenance pH sensor combined with a PT100 temperature sensor for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.