Learn More
METTLER TOLEDO pH Sensor InPro3253i/SG/120
Supplier:  METTLER TOLEDO 52005377
Encompass
The InPro 3253i SG/120 is a digital, pre-pressurized, low maintenance ISM pH & ORP sensor combined with a temperature sensor for in-line measurement in demanding applications. The 120 mm version is optimal for short installation lengths.
Catalog No. NC0104921