Learn More
METTLER TOLEDO pH Sensor InPro4260i/SG/120
Supplier:  METTLER TOLEDO 52005381
Encompass
The InPro 4260i is a combined pH and ORP (redox) probe with integrated temperature sensor designed for use in highly demanding chemical applications. It provides ISM advanced diagnostics to optimize maintenance decisions and sensor lifetime.
Catalog No. NC1042522