Learn More
METTLER TOLEDO DO Sensor InPro6830/12/120
Catalog No. NC0648631 Shop All METTLER TOLEDO Products
Encompass
The InPro 6830 dissolved oxygen sensor with 12 mm diameter body provides maximum accuracy and ultimate cleanability for vessels with limited space or in containers with smaller volumes.
Catalog No. NC0648631 Supplier METTLER TOLEDO Supplier No. 52201016
The InPro 6830 dissolved oxygen sensor with 12 mm diameter body provides maximum accuracy and ultimate cleanability for vessels with limited space or in containers with smaller volumes.