Learn More
METTLER TOLEDO pH Sensor InPro 3253i SG/325
Catalog No. NC0214263 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253i SG/325 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 325 mm version is optimal for medium to long installation lengths.
Catalog No. NC0214263 Supplier METTLER TOLEDO Supplier No. 52005379
The InPro 3253i SG/325 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 325 mm version is optimal for medium to long installation lengths.