Learn More
METTLER TOLEDO pH Sensor InPro3100/120/PT1000
Supplier:  METTLER TOLEDO 52000658
Encompass
The InPro 3100 is a pH electrode with temperature sensor, designed for measurement in bioprocesses where CIP and SIP procedures are required. The 120 mm version is optimal for short installation lengths.
Catalog No. NC9799323