Learn More
METTLER TOLEDO pH Sensor InPro 3250SG 225/PT100
Catalog No. NC9876486 Shop All METTLER TOLEDO Products
Encompass
The InPro 3250SG 225/PT100 is a pre-pressurized, liquid filled, low maintenance pH and ORP (Redox) sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. This version is 225mm long.
Catalog No. NC9876486 Supplier METTLER TOLEDO Supplier No. 52002560
The InPro 3250SG 225/PT100 is a pre-pressurized, liquid filled, low maintenance pH and ORP (Redox) sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. This version is 225mm long.