Learn More
METTLER TOLEDO pH Sensor InPro 3253SG 225/PT1000
Catalog No. NC0214262 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253SG 225/PT1000 is a pre-pressurized, liquid filled, low maintenance pH & ORP (Redox) sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. This version is 225 mm long.
Catalog No. NC0214262 Supplier METTLER TOLEDO Supplier No. 52002579
The InPro 3253SG 225/PT1000 is a pre-pressurized, liquid filled, low maintenance pH & ORP (Redox) sensor, combined with a PT1000 temperature sensor, for in-line measurement in demanding applications. This version is 225 mm long.