Learn More
METTLER TOLEDO InPro3253 / SG / 325 / PT1000 pH sensor
Catalog No. NC1180648 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253SG 325 / PT1000 is a pre-pressurized, liquid-filled, low-maintenance pH / ORP (Redox) sensor combined with a PT1000 temperature sensor for in-line measurement for demanding applications. This version has a length of 325.
Catalog No. NC1180648 Supplier METTLER TOLEDO Supplier No. 52002581
The InPro 3253SG 325 / PT1000 is a pre-pressurized, liquid-filled, low-maintenance pH / ORP (Redox) sensor combined with a PT1000 temperature sensor for in-line measurement for demanding applications. This version has a length of 325.