Learn More
Cayman Chemical InRleukIn6RaCD126 RabbI 100uL
SDP
Catalog No. 502916067
Encompass_Preferred
An agonist of NPY receptor Y5 inhibits forskolin-induced cAMP production in HEK293 cells expressing the Y5 receptor (EC50 17 nM) selectively binds to Y5 (IC50 0.24 nM in HEK293 cells expressing the human receptor) over Y1 Y2 and Y4 (IC50s 530 500 and 51 nM respectively in BHK-21 cells expressing the human receptors) increases VEGF levels as well as endothelial branch length and branch points in 4T1 cells in a concentration-dependent manner induces relaxation of vasopressin-precontracted rat mesenteric arteries at 1 UM increases food intake in rats at 0.2 and 2 nmol/animal i.c.v.
Catalog No. 50-291-6067 Supplier Cayman Chemical Supplier No. 38089100 UL
An agonist of NPY receptor Y5 inhibits forskolin-induced cAMP production in HEK293 cells expressing the Y5 receptor (EC50 17 nM) selectively binds to Y5 (IC50 0.24 nM in HEK293 cells expressing the human receptor) over Y1 Y2 and Y4 (IC50s 530 500 and 51 nM respectively in BHK-21 cells expressing the human receptors) increases VEGF levels as well as endothelial branch length and branch points in 4T1 cells in a concentration-dependent manner induces relaxation of vasopressin-precontracted rat mesenteric arteries at 1 UM increases food intake in rats at 0.2 and 2 nmol/animal i.c.v.