Learn More
Echelon Biosciences Research Labs Ins(1,3,4,5)P4 BIOTIN 10ug
SDP
Catalog No. 50143994
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. This Inositol 1,3,4,5-tetrakisphosphate (Ins(1,3,4,5)P4) analog is labeled with biotin through a three-carbon linker at the 1-position on the inositol ring. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-143-994 Supplier Echelon Biosciences Research Labs Supplier No. Q139B10UG
Inositol phosphates are important signaling molecules involved in numerous cellular functions. This Inositol 1,3,4,5-tetrakisphosphate (Ins(1,3,4,5)P4) analog is labeled with biotin through a three-carbon linker at the 1-position on the inositol ring. This item is non returnable