Learn More
Echelon Biosciences Research Labs Ins(1,3,4,5)P4 BIOTIN 50ug
SDP
Supplier:  Echelon Biosciences Research Labs Q139B50UG
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. This Inositol 1,3,4,5-tetrakisphosphate (Ins(1,3,4,5)P4) analog is labeled with biotin through a three-carbon linker at the 1-position on the inositol ring. This item is non returnable
Catalog No. 50-143-995