Learn More
Echelon Biosciences Research Labs Ins(1,3,4)P3 500 ug
SDP
Catalog No. 50720723
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,3,4-trisphosphate (Ins(1,3,4)P3) is an intermediate in the inositol phosphate pathway. Ins(1,3,4)P3 inhibits Ins(3,4,5,6)P4 1-kinase which regulates Ins(3,4,5,6)P4 levels. Ins(3,4,5,6)P4 in turn is responsible for inhibiting calcium-activated chloride channels. Ins(1,3,4)P3 is produced by dephosphorylation of Ins(1,3,4,5)P4 by a 5-phosphatase. Ins(1,3,4)P3 is a substrate for phosphatases yielding Ins(1,3)P2 and Ins(3,4)P2. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-723 Supplier Echelon Biosciences Research Labs Supplier No. Q0134500UG
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,3,4-trisphosphate (Ins(1,3,4)P3) is an intermediate in the inositol phosphate pathway. Ins(1,3,4)P3 inhibits Ins(3,4,5,6)P4 1-kinase which regulates Ins(3,4,5,6)P4 levels. Ins(3,4,5,6)P4 in turn is responsible for inhibiting calcium-activated chloride channels. Ins(1,3,4)P3 is produced by dephosphorylation of Ins(1,3,4,5)P4 by a 5-phosphatase. Ins(1,3,4)P3 is a substrate for phosphatases yielding Ins(1,3)P2 and Ins(3,4)P2. This item is non returnable