Learn More
Echelon Biosciences Research Labs Ins(1,3,5)P3 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs Q0135100UG
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,3,5-trisphosphate (Ins(1,3,5)P3) is not an intermediate in the inositol phosphate pathway but corresponds to the PtdIns(3,5)P2 headgroup. This item is non returnable
Catalog No. 50-720-725