Learn More
Echelon Biosciences Research Labs Ins(1,4,5)P3 500 ug
SDP
Catalog No. 50720729
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3), aka IP3, is a second messenger produced by phospholipase C catalyzed hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2). IP3 binds to the IP3-receptor on the endoplasmic reticulum causing calcium to be released through the opening of a tetrameric ion channel. IP3 is rapidly metabolized by a specific series of inositol phosphatases and kinases. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-729 Supplier Echelon Biosciences Research Labs Supplier No. Q0145500UG
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3), aka IP3, is a second messenger produced by phospholipase C catalyzed hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2). IP3 binds to the IP3-receptor on the endoplasmic reticulum causing calcium to be released through the opening of a tetrameric ion channel. IP3 is rapidly metabolized by a specific series of inositol phosphatases and kinases. This item is non returnable