Learn More
Echelon Biosciences Research Labs Ins(1,5)P2 1 mg
SDP
Catalog No. 50720712
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,5-bisphosphate (Ins(1,5)P2) is not an intermediate in the inositol phosphate pathway but corresponds to the PtdIns(5)P headgroup. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-712 Supplier Echelon Biosciences Research Labs Supplier No. Q00151MG
Add to Cart
Add to Cart
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,5-bisphosphate (Ins(1,5)P2) is not an intermediate in the inositol phosphate pathway but corresponds to the PtdIns(5)P headgroup. This item is non returnable