Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ INS-1 832/3 Rat Insulinoma Cell Line

Catalog No. SCC208 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
≥ 1 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC208 ≥ 1 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC208 Supplier MilliporeSigma™ Supplier No. SCC208
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

INS-1 832/3 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.

  • Glucose-stimulated insulin secretion (GSIS) is potentiated by pancreatic beta cells and is critical to the physiological control of blood glucose levels
  • Insulin secretion is impaired in type 2 diabetes, and insight into the mechanisms and regulation of insulin secretion is fundamental to understanding the roles of beta cells in metabolic disease
  • The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality
  • Cells are tested negative for infectious diseases by a Mouse/Rat  Comprehensive CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are verified to be of rat origin and negative for inter-species contamination from mouse, Chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
Format Vial
Quantity ≥ 1 x 106 cells/vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.