Learn More
ADDEXBIO TECHNOLOGIES INS-1 Cell Line, Catalog Number C0018007, Insulinoma, Derived from Rattus Norvegicus Pancreatic Islets, 5% CO2, 37°C, Biosafety Level I
Catalog No. NC0898977
Encompass
INS-1 Cell Line, Catalog Number C0018007, Insulinoma, Derived from Rattus Norvegicus Pancreatic Islets, 5% CO2, 37°C, Biosafety Level I
Catalog No. NC0898977 Supplier ADDEXBIO TECHNOLOGIES Supplier No. C0018007
INS-1 Cell Line, Catalog Number C0018007, Insulinoma, Derived from Rattus Norvegicus Pancreatic Islets, 5% CO2, 37°C, Biosafety Level I