Learn More
MSC Polar Tech Cooler, 15 in, 2 in Wall, 1 ea
Catalog No. 501920780
Encompass
This insulated shipping box is designed to maintain the integrity of temperature-sensitive products during transport. It features a high-quality foam block interior to insulate contents like lab samples and chemicals, and a durable cardboard exterior to withstand shipping conditions.
  • Maintains insulation and integrity of temperature-sensitive products
  • Economical, lightweight, reusable, and recyclable construction
  • Thick molded EPS foam container protects contents from crushing and breakage
  • Cartons are printed with 'Rush, Perishable, and This Side Up' shipping instructions
  • Includes a seamless molded EPS foam body with a tight-fitting lid and a 200#/ECT-32 white corrugated carton
Catalog No. 50-192-0780 Supplier MSC Supplier No. 39559299
This insulated shipping box is designed to maintain the integrity of temperature-sensitive products during transport. It features a high-quality foam block interior to insulate contents like lab samples and chemicals, and a durable cardboard exterior to withstand shipping conditions.
  • Maintains insulation and integrity of temperature-sensitive products
  • Economical, lightweight, reusable, and recyclable construction
  • Thick molded EPS foam container protects contents from crushing and breakage
  • Cartons are printed with 'Rush, Perishable, and This Side Up' shipping instructions
  • Includes a seamless molded EPS foam body with a tight-fitting lid and a 200#/ECT-32 white corrugated carton