Learn More
Cayman Chemical inS3-54-A18, Purity: ≥98%, CAS: 328998-53-4, 25 mg
SDP
Catalog No. 502914733
Encompass_Preferred
A STAT3 inhibitor; selectively inhibits the binding of STAT3 to DNA over STAT1 at 25 µM; inhibits STAT3 activity in a reporter assay using MDA-MB-231 cells at 20 µM; induces apoptosis in, and reduces migration and invasion of, A549 and MDA-MB-231 cells at 5 or 10 µM; reduces tumor volume in an A549 mouse xenograft model at 200 mg/kg
Catalog No. 50-291-4733 Supplier Cayman Chemical Supplier No. 3967825 MG
A STAT3 inhibitor; selectively inhibits the binding of STAT3 to DNA over STAT1 at 25 µM; inhibits STAT3 activity in a reporter assay using MDA-MB-231 cells at 20 µM; induces apoptosis in, and reduces migration and invasion of, A549 and MDA-MB-231 cells at 5 or 10 µM; reduces tumor volume in an A549 mouse xenograft model at 200 mg/kg