Learn More
TARGETMOL CHEMICALS INC INS3-54-A26 5MG
SDP
Catalog No. 502580598
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. inS3-54-A26 is an inhibitor of the DNA-binding domain of STAT3. inS3-54-A26 suppresses tumor growth metastasis and the gene expression of STAT3. purity: 98%
Catalog No. 50-258-0598 Supplier TARGETMOL CHEMICALS INC Supplier No. T276135MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. inS3-54-A26 is an inhibitor of the DNA-binding domain of STAT3. inS3-54-A26 suppresses tumor growth metastasis and the gene expression of STAT3. purity: 98%