Learn More
Alkali Scientific ASI™ Insect Breeding Dish, PS, 50x15mm, ventilation mesh 3.2mm pore 0.053μm, 20 sleeve, 200 per Case
SDP
Catalog No. 503028681
Encompass_Preferred
Insect Breeding Dish, PS, 50x15mm, ventilation hole 13.2mm   20 sleeve, 200 per Case
Catalog No. 50-302-8681 Supplier Alkali Scientific Supplier No. LP350
Insect Breeding Dish, PS, 50x15mm, ventilation hole 13.2mm   20 sleeve, 200 per Case