Learn More
Discovery Life Sciences INSECT CELL CONTROL MICROSOMES
SDP
Catalog No. NC9892378
Encompass
Insect Cell Control SUPERSOMES 1.5mL Insect cells (BTI-TN-5B1-4) infected with wild type baculovirus (Autographa californica) were used to prepare these microsomes
Catalog No. NC9892378 Supplier Discovery Life Sciences Supplier No. 456200
Insect Cell Control SUPERSOMES 1.5mL Insect cells (BTI-TN-5B1-4) infected with wild type baculovirus (Autographa californica) were used to prepare these microsomes