Learn More
Discovery Life Sciences INSECT CELL CONTROL MICROSOMES
SDP
Supplier:  Discovery Life Sciences 456201
Encompass
Insect Cell Control SUPERSOMES 0.5mL Insect cells (BTI-TN-5B1-4) infected with wild type baculovirus (Autographa californica) were used to prepare these microsomes
Catalog No. NC9496057