Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Insect Masks
SDPSCI_ED

Catalog No. S13233
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13233 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13233 Supplier Roylco™ Supplier No. R52078
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

24 card-stock masks printed on ethically-sourced card with vegetable based ink.

  • Color with crayon, paint and markers
  • Add chencille stems to hold the masks in place
  • Research each insect and act out dramatic scenes or recreate 'real' behaviors
  • 8 different designs, up to 20 x 25 cm (8 x 10 in.)

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Quantity 24 Masks
Languages English, Spanish, French, German
Subject Biology - Insects
Length (English) 10 in.
Length (Metric) 25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.