Learn More
Fine Science Tools Insect Pins, Tip: 0.03 mm, 4 cm, 100/pk
SDP
Catalog No. NC0167081
Encompass
Insect Pins - Stainless Steel/Size 00/ Size: 00/ Tip Diameter: 0.03mm/ Rod Diameter: 0.3mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100
Catalog No. NC0167081 Supplier Fine Science Tools Supplier No. 2600130
Insect Pins - Stainless Steel/Size 00/ Size: 00/ Tip Diameter: 0.03mm/ Rod Diameter: 0.3mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100