Learn More
Fine Science Tools Insect Pins, Tip: 0.03 mm, 4 cm, 100/pk
SDP
Catalog No. NC0167082
Encompass
Insect Pins - Stainless Steel/Size 5/ Size: 5/ Tip Diameter: 0.03mm/ Rod Diameter: 0.6mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100
Catalog No. NC0167082 Supplier Fine Science Tools Supplier No. 2600160
Insect Pins - Stainless Steel/Size 5/ Size: 5/ Tip Diameter: 0.03mm/ Rod Diameter: 0.6mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100