Learn More
Caron Products And Services Insect Rearing Chamber, 33 cu. ft., Temp, No RH
Supplier:  Caron Products And Services 7340332
Encompass_Preferred
Carons 33 cu. ft. (934 liters) Insect Rearing Chamber. Includes touchscreen controls, temperature range from 10C to 40C, gROD Refrigeration on Demand, and electrostatically coated refrigeration coils. Provides high volume solution for a wide variety of entomological studies. 230V, 60 Hz.
Catalog No. NC2676813
Add to Cart
Request a Quote