Learn More
Caron Products And Services Insect Rearing Chamber, 33 cu. ft., Temp, No RH
Catalog No. NC2676812
Encompass_Preferred
Carons 33 cu. ft. (934 liters) Insect Rearing Chamber. Includes touchscreen controls, temperature range from 10C to 40C, gROD Refrigeration on Demand, and electrostatically coated refrigeration coils. Provides high volume solution for a wide variety of entomological studies. 115V, 60 Hz.
Catalog No. NC2676812 Supplier Caron Products And Services Supplier No. 7340331
Add to Cart
Add to Cart
Request a Quote
Carons 33 cu. ft. (934 liters) Insect Rearing Chamber. Includes touchscreen controls, temperature range from 10C to 40C, gROD Refrigeration on Demand, and electrostatically coated refrigeration coils. Provides high volume solution for a wide variety of entomological studies. 115V, 60 Hz.