Learn More
Medchemexpress LLC Insect repellent M 3 10mM 1mL
SDP
Catalog No. 5000205593
Encompass_Preferred
Insect repellent M 3535 (Ethyl butylacetylaminopropionate IR-3535) is a potent anthelmintic that is toxic to zebrafish with a lethal concentration 50 (LC50) of 140 mg/L at 72 hours post fertilization (hpf) Insect repellent M 3535 causes morphological abnormalities and cardiac defects in early embryonic development of zebrafish by potentially inducing the production and accumulation of reactive oxygen species (Reactive Oxygen Species) and activating oxidative stress responses thereby activating the endogenous apoptosis pathway[1]
Catalog No. 50-002-05593 Supplier Medchemexpress LLC Supplier No. HYW09947910MM/1ML
Insect repellent M 3535 (Ethyl butylacetylaminopropionate IR-3535) is a potent anthelmintic that is toxic to zebrafish with a lethal concentration 50 (LC50) of 140 mg/L at 72 hours post fertilization (hpf) Insect repellent M 3535 causes morphological abnormalities and cardiac defects in early embryonic development of zebrafish by potentially inducing the production and accumulation of reactive oxygen species (Reactive Oxygen Species) and activating oxidative stress responses thereby activating the endogenous apoptosis pathway[1]