Learn More
Lonza Walkersville Insect-XPRESS Insect Cell Med
Catalog No. 5000496404
Encompass
Insect-XPRESS Medium is a protein-free formulation designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21) and Drosophila melanogaster (S2) Cell densities in excess of 8 3 10 6 cells/ml can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and an excess of oxygen This formulation can also be used for stationary monolayer cultures and shake-flask cultures Insect-XPRESS Medium contains L-glutamine and supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS) For cryopreservation of insect cells Insect-XPRESS can be mixed 50 50 with Cryoprotective Medium (Cat No 12-769E) All BioWhittaker Cell Culture Media products are for Research Use Only (RUO) and are not approved for human or veterinary use or for use in clinical or in vitro diagnostic procedures
Catalog No. 50-004-96404 Supplier Lonza Walkersville Supplier No. 12730Q
Add to Cart
Add to Cart
Insect-XPRESS Medium is a protein-free formulation designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21) and Drosophila melanogaster (S2) Cell densities in excess of 8 3 10 6 cells/ml can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and an excess of oxygen This formulation can also be used for stationary monolayer cultures and shake-flask cultures Insect-XPRESS Medium contains L-glutamine and supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS) For cryopreservation of insect cells Insect-XPRESS can be mixed 50 50 with Cryoprotective Medium (Cat No 12-769E) All BioWhittaker Cell Culture Media products are for Research Use Only (RUO) and are not approved for human or veterinary use or for use in clinical or in vitro diagnostic procedures